Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sphkap
Entrez Gene ID
316561
Gene Name
SPHK1 interactor, AKAP domain containing
Gene Aliases
RGD1311951, Skip
Chromosome Location
Chr.9: 89050405 - 89195445 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127492.1 NP_001120964.1 2 - 3 260 65
XM_006245230.2 XP_006245292.1 2 - 3 432 65
XM_006245231.2 XP_006245293.1 2 - 3 833 65
XM_006245234.2 XP_006245296.1 2 - 3 431 65

View Full Product Details