Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mogat1
Entrez Gene ID
363261
Gene Name
monoacylglycerol O-acyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.9: 84410972 - 84418522 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108803.1 NP_001102273.1 1 - 2 187 80

View Full Product Details