Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnkd
Entrez Gene ID
100188944
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
Mr-1
Chromosome Location
Chr.9: 81566074 - 81634531 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134750.1 NP_001128222.1 5 - 6 529 70
NM_001134751.1 NP_001128223.1 4 - 5 447 70
GenBank mRNA BC168228.1 - 5 - 6 529 70

View Full Product Details