Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf25
Entrez Gene ID
301515
Gene Name
ring finger protein 25
Gene Aliases
-
Chromosome Location
Chr.9: 81873293 - 81880136 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012004.2 NP_001012004.2 4 - 5 297 66
XM_006245172.2 XP_006245234.1 4 - 5 328 66
XM_006245173.1 XP_006245235.1 4 - 5 1178 66
GenBank mRNA CF114855.1 - 4 - 5 307 66

View Full Product Details