The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012004.2 | NP_001012004.2 | 8 - 9 | 661 | 81 |
XM_006245172.2 | XP_006245234.1 | 8 - 9 | 689 | 81 | |
XM_006245173.1 | XP_006245235.1 | 8 - 9 | 1542 | 81 | |
GenBank mRNA | CB691642.1 | - | 4 - 5 | 330 | 81 |