Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cobl
Entrez Gene ID
305497
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
-
Chromosome Location
Chr.14: 92341949 - 92497173 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107236.1 NP_001100706.1 6 - 7 993 74
XM_008770308.1 XP_008768530.1 6 - 7 2271 74
XM_008770309.1 XP_008768531.1 5 - 6 2196 74
XM_008770310.1 XP_008768532.1 5 - 6 2195 74
XM_008770311.1 XP_008768533.1 5 - 6 2197 74

View Full Product Details