Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map2
Entrez Gene ID
25595
Gene Name
microtubule-associated protein 2
Gene Aliases
MAP2R, Mtap2
Chromosome Location
Chr.9: 73334908 - 73462972 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245100.2 XP_006245162.1 11 - 12 9407 106
XM_008767197.1 XP_008765419.1 11 - 12 5438 106
XM_008767198.1 XP_008765420.1 11 - 12 5438 106
XM_008767199.1 XP_008765421.1 11 - 12 5435 106
XM_008767200.1 XP_008765422.1 11 - 12 5435 106
XM_008767201.1 XP_008765423.1 11 - 12 5435 106
XM_008767202.1 XP_008765424.1 11 - 12 5426 106
XM_008767204.1 XP_008765426.1 10 - 11 5192 106
XM_008767205.1 XP_008765427.1 10 - 11 5180 106
XM_008767207.1 XP_008765429.1 9 - 10 1245 106
GenBank mRNA BC081835.1 - 9 - 10 1256 106
X74211.1 - 8 - 9 1131 106

View Full Product Details