Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc27a5
Entrez Gene ID
79111
Gene Name
solute carrier family 27 (fatty acid transporter), member 5
Gene Aliases
BAL, rBAL-1
Chromosome Location
Chr.1: 65576599 - 65587192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024143.2 NP_077057.1 1 - 2 735 99
GenBank mRNA AF242189.1 - 1 - 2 736 99
BC091147.1 - 1 - 2 758 99

View Full Product Details