The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024143.2 | NP_077057.1 | 9 - 10 | 1941 | 76 |
GenBank mRNA | AF242189.1 | - | 9 - 10 | 1942 | 76 |
BC091147.1 | - | 9 - 10 | 1964 | 76 |