Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map7d3
Entrez Gene ID
317614
Gene Name
MAP7 domain containing 3
Gene Aliases
RGD1565514
Chromosome Location
Chr.X: 159176825 - 159220494 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271327.1 NP_001258256.1 8 - 9 1430 57
XM_008773649.1 XP_008771871.1 8 - 9 1602 57
XM_008773650.1 XP_008771872.1 8 - 9 1514 57

View Full Product Details