Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cadm2
Entrez Gene ID
360687
Gene Name
cell adhesion molecule 2
Gene Aliases
Igsf4d, Necl-3, SynCAM 2
Chromosome Location
Chr.11: 4175078 - 4397335 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047102.1 NP_001040567.1 2 - 3 360 95
GenBank mRNA DQ272744.1 - 2 - 3 360 95

View Full Product Details