The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100658.2 | NP_001094128.2 | 9 - 10 | 1073 | 85 |
GenBank mRNA | S75280.1 | - | 9 - 10 | 1020 | 85 |