The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106413.1 | NP_001099883.1 | 1 - 2 | 361 | 71 |
XM_008760760.1 | XP_008758982.1 | 1 - 2 | 488 | 71 | |
XM_008760761.1 | XP_008758983.1 | 2 - 3 | 411 | 71 |