Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Card6
Entrez Gene ID
294770
Gene Name
caspase recruitment domain family, member 6
Gene Aliases
-
Chromosome Location
Chr.2: 54810676 - 54824044 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106413.1 NP_001099883.1 3 - 4 3130 81

View Full Product Details