Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr70
Entrez Gene ID
294783
Gene Name
WD repeat domain 70
Gene Aliases
RGD1309487
Chromosome Location
Chr.2: 56944462 - 57196522 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013909.1 NP_001013931.1 14 - 15 1530 79
GenBank mRNA BC089903.1 - 14 - 15 1530 79

View Full Product Details