Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csgalnact1
Entrez Gene ID
306375
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
Chgn, RGD1307618
Chromosome Location
Chr.16: 22704412 - 23075071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107309.1 NP_001100779.1 1 - 2 635 82
XM_006253026.2 XP_006253088.1 2 - 3 1091 82
XM_006253027.2 XP_006253089.1 3 - 4 1186 82
XM_006253028.2 XP_006253090.1 3 - 4 1600 82
XM_006253029.2 XP_006253091.1 3 - 4 1406 82
XM_006253030.2 XP_006253092.1 3 - 4 1490 82
XM_006253031.2 XP_006253093.1 2 - 3 1436 82
XM_008771110.1 XP_008769332.1 3 - 4 1220 82
XM_008771111.1 XP_008769333.1 3 - 4 1225 82
XM_008771112.1 XP_008769334.1 3 - 4 1226 82
XM_008771113.1 XP_008769335.1 3 - 4 1225 82
XM_008771114.1 XP_008769336.1 3 - 4 1224 82
XM_008771115.1 XP_008769337.1 5 - 6 1360 82
XM_008771116.1 XP_008769338.1 3 - 4 1364 82
GenBank mRNA BC168857.1 - 1 - 2 859 82

View Full Product Details