Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csgalnact1
Entrez Gene ID
306375
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
Chgn, RGD1307618
Chromosome Location
Chr.16: 22704412 - 23075071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107309.1 NP_001100779.1 5 - 6 1217 61
XM_006253026.2 XP_006253088.1 6 - 7 1673 61
XM_006253027.2 XP_006253089.1 7 - 8 1768 61
XM_006253028.2 XP_006253090.1 7 - 8 2182 61
XM_006253029.2 XP_006253091.1 7 - 8 1988 61
XM_006253030.2 XP_006253092.1 7 - 8 2072 61
XM_006253031.2 XP_006253093.1 6 - 7 2018 61
XM_008771110.1 XP_008769332.1 7 - 8 1802 61
XM_008771111.1 XP_008769333.1 7 - 8 1807 61
XM_008771112.1 XP_008769334.1 7 - 8 1808 61
XM_008771113.1 XP_008769335.1 7 - 8 1807 61
XM_008771114.1 XP_008769336.1 7 - 8 1806 61
XM_008771115.1 XP_008769337.1 9 - 10 1942 61
XM_008771116.1 XP_008769338.1 7 - 8 1946 61
GenBank mRNA BC168857.1 - 5 - 6 1441 61

View Full Product Details