The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013121.1 | NP_001013139.1 | 4 - 5 | 774 | 61 |
XM_008768956.1 | XP_008767178.1 | 4 - 5 | 877 | 61 | |
GenBank mRNA | BC079077.1 | - | 4 - 5 | 774 | 61 |
FQ214278.1 | - | 3 - 4 | 604 | 61 |