The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037203.1 | NP_001032280.1 | 5 - 6 | 504 | 101 |
XM_006248766.2 | XP_006248828.1 | 5 - 6 | 614 | 101 | |
GenBank mRNA | BC107945.1 | - | 5 - 6 | 504 | 101 |