The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100547.1 | NP_001094017.1 | 1 - 2 | 519 | 86 |
GenBank mRNA | X94186.1 | - | 1 - 2 | 102 | 86 |