Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mon1a
Entrez Gene ID
315999
Gene Name
MON1 secretory trafficking family member A
Gene Aliases
RGD1308823
Chromosome Location
Chr.8: 116657348 - 116682172 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126284.1 NP_001119756.1 5 - 6 1696 63
XM_006243782.1 XP_006243844.1 5 - 6 1585 63
XM_006243783.2 XP_006243845.1 5 - 6 1830 63
XM_008766536.1 XP_008764758.1 4 - 5 1345 63
GenBank mRNA BC161958.1 - 5 - 6 1715 63

View Full Product Details