Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map4
Entrez Gene ID
367171
Gene Name
microtubule-associated protein 4
Gene Aliases
Mtap4
Chromosome Location
Chr.8: 118013586 - 118159934 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024278.1 NP_001019449.1 9 - 10 2150 67
XM_006243813.2 XP_006243875.1 10 - 11 5402 67
XM_006243814.2 XP_006243876.1 10 - 11 5402 67
XM_006243815.2 XP_006243877.1 10 - 11 5402 67
XM_006243816.2 XP_006243878.1 10 - 11 5402 67
XM_006243818.2 XP_006243880.1 10 - 11 5403 67
XM_006243819.2 XP_006243881.1 10 - 11 5403 67
XM_006243821.2 XP_006243883.1 9 - 10 2158 67
XM_006243822.2 XP_006243884.1 9 - 10 2158 67
XM_006243823.2 XP_006243885.1 9 - 10 2158 67
GenBank mRNA AB175782.1 - 1 - 2 68 67
BC088405.1 - 9 - 10 2150 67

View Full Product Details