The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191796.1 | NP_001178725.1 | 3 - 4 | 370 | 107 |
XM_008766293.1 | XP_008764515.1 | 3 - 4 | 587 | 107 |