Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tma7
Entrez Gene ID
679747
Gene Name
translational machinery associated 7 homolog
Gene Aliases
Ccdc72
Chromosome Location
Chr.8: 117815725 - 117820413 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126048.2 NP_001119520.1 3 - 4 227 118
GenBank mRNA BC157813.1 - 3 - 4 227 118
FQ221914.1 - 3 - 4 118
FQ222972.1 - 3 - 4 192 118

Gene Symbol
LOC100910678
Entrez Gene ID
100910678
Gene Name
coiled-coil domain-containing protein 72-like
Gene Aliases
-
Chromosome Location
Chr.4: 79015058 - 79015706 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749743.3 XP_003749791.1 2 - 2 203 118
XM_006236476.2 XP_006236538.1 1 - 1 355 118

View Full Product Details