The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022503.2 | NP_071948.1 | 3 - 4 | 278 | 78 |
GenBank mRNA | AW521400.1 | - | 1 - 2 | 78 | |
BC166880.1 | - | 3 - 4 | 246 | 78 | |
BG668415.1 | - | 3 - 4 | 285 | 78 | |
FQ209801.1 | - | 3 - 4 | 273 | 78 | |
FQ209847.1 | - | 3 - 4 | 245 | 78 | |
FQ210551.1 | - | 3 - 4 | 245 | 78 | |
FQ211276.1 | - | 3 - 4 | 246 | 78 | |
FQ214317.1 | - | 3 - 4 | 254 | 78 | |
FQ217361.1 | - | 3 - 4 | 245 | 78 | |
FQ221839.1 | - | 3 - 4 | 250 | 78 | |
FQ223968.1 | - | 3 - 4 | 269 | 78 | |
FQ228537.1 | - | 3 - 4 | 317 | 78 | |
FQ229029.1 | - | 3 - 4 | 247 | 78 | |
FQ229035.1 | - | 3 - 4 | 279 | 78 | |
X54080.1 | - | 3 - 4 | 252 | 78 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751367.3 | XP_003751415.1 | 1 - 1 | 229 | 78 |