The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107084.1 | NP_001100554.1 | 3 - 4 | 354 | 64 |
XM_008768799.1 | XP_008767021.1 | 2 - 3 | 1227 | 64 | |
GenBank mRNA | BC166425.1 | - | 3 - 4 | 254 | 64 |