Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcl6
Entrez Gene ID
303836
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
-
Chromosome Location
Chr.11: 80255722 - 80279077 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107084.1 NP_001100554.1 4 - 5 577 87
XM_008768799.1 XP_008767021.1 3 - 4 1450 87
GenBank mRNA BC166425.1 - 4 - 5 477 87

View Full Product Details