Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl6
Entrez Gene ID
287974
Gene Name
kelch-like family member 6
Gene Aliases
-
Chromosome Location
Chr.11: 84745958 - 84785315 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105867.1 NP_001099337.1 5 - 6 1376 66
XM_006248594.1 XP_006248656.1 5 - 6 1147 66

View Full Product Details