Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sdf2l1
Entrez Gene ID
680945
Gene Name
stromal cell-derived factor 2-like 1
Gene Aliases
-
Chromosome Location
Chr.11: 88122271 - 88124513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109433.1 NP_001102903.1 1 - 2 240 59
GenBank mRNA FQ225991.1 - 1 - 2 216 59
FQ230956.1 - 1 - 2 217 59

View Full Product Details