Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tp63
Entrez Gene ID
246334
Gene Name
tumor protein p63
Gene Aliases
Ket, P73l, Tp73l, Trp63
Chromosome Location
Chr.11: 78234853 - 78456329 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127339.1 NP_001120811.1 5 - 6 912 99
NM_001127341.1 NP_001120813.1 5 - 6 912 99
NM_001127342.1 NP_001120814.1 3 - 4 483 99
NM_001127343.1 NP_001120815.1 3 - 4 483 99
NM_001127344.1 NP_001120816.1 3 - 4 483 99
NM_019221.3 NP_062094.1 5 - 6 912 99
XM_006248500.2 XP_006248562.1 5 - 6 993 99
XM_008768791.1 XP_008767013.1 6 - 7 842 99
GenBank mRNA AJ277446.1 - 4 - 5 714 99
AJ277447.1 - 3 - 4 483 99
AJ277448.1 - 4 - 5 714 99
AJ277449.1 - 5 - 6 765 99
AJ277450.1 - 3 - 4 483 99
AJ277451.1 - 5 - 6 765 99
AJ277452.1 - 4 - 5 714 99
AJ277453.1 - 3 - 4 483 99
Y10258.2 - 5 - 6 912 99

View Full Product Details