The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039021.1 | NP_001034110.1 | 10 - 11 | 2247 | 77 |
GenBank mRNA | AY871204.1 | - | 10 - 11 | 2247 | 77 |