The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024865.1 | NP_001020036.1 | 3 - 4 | 378 | 102 |
GenBank mRNA | BC097472.1 | - | 3 - 4 | 378 | 102 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006252309.2 | XP_006252371.1 | 1 - 1 | 162 | 102 |