The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014148.1 | NP_001014170.1 | 5 - 6 | 646 | 70 |
XM_008772044.1 | XP_008770266.1 | 3 - 4 | 493 | 70 | |
XM_008772046.1 | XP_008770268.1 | 3 - 4 | 527 | 70 | |
XM_008772047.1 | XP_008770269.1 | 4 - 5 | 549 | 70 | |
XM_008772048.1 | XP_008770270.1 | 4 - 5 | 566 | 70 | |
XM_008772049.1 | XP_008770271.1 | 3 - 4 | 639 | 70 | |
GenBank mRNA | BC079261.1 | - | 5 - 6 | 646 | 70 |