Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a46
Entrez Gene ID
291709
Gene Name
solute carrier family 25, member 46
Gene Aliases
RGD1305072
Chromosome Location
Chr.18: 24369104 - 24397412 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100515.1 NP_001093985.1 4 - 5 521 99
GenBank mRNA BC166810.1 - 4 - 5 521 99

View Full Product Details