Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rprd1a
Entrez Gene ID
291736
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
Gene Aliases
P15rs, RGD1307374
Chromosome Location
Chr.18: 16450163 - 16497910 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772003.1 XP_008770225.1 2 - 3 433 65
XM_214609.8 XP_214609.6 2 - 3 433 65

View Full Product Details