Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf14
Entrez Gene ID
619577
Gene Name
ring finger protein 14
Gene Aliases
ARA54
Chromosome Location
Chr.18: 31443432 - 31468641 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034995.1 NP_001030167.1 6 - 7 1318 135
XM_006254636.2 XP_006254698.1 6 - 7 1412 135
XM_006254637.2 XP_006254699.1 6 - 7 1726 135
XM_006254639.2 XP_006254701.1 7 - 8 1523 135
XM_006254640.2 XP_006254702.1 5 - 6 1262 135
XM_006254641.2 XP_006254703.1 4 - 5 928 135
XM_008772069.1 XP_008770291.1 6 - 7 1278 135
GenBank mRNA BC103648.1 - 6 - 7 1318 135
BC166497.1 - 3 - 4 789 135

View Full Product Details