Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mocos
Entrez Gene ID
361300
Gene Name
molybdenum cofactor sulfurase
Gene Aliases
-
Chromosome Location
Chr.18: 16590100 - 16636084 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108425.1 NP_001101895.1 11 - 12 2366 85
XM_006254466.2 XP_006254528.1 10 - 11 2533 85

View Full Product Details