Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stard4
Entrez Gene ID
291699
Gene Name
StAR-related lipid transfer (START) domain containing 4
Gene Aliases
-
Chromosome Location
Chr.18: 25984638 - 25997555 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106159.2 NP_001099629.1 5 - 6 568 89

View Full Product Details