The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012163.1 | NP_001012163.1 | 7 - 8 | 929 | 76 |
XM_006254596.2 | XP_006254658.1 | 7 - 8 | 1066 | 76 | |
XM_006254597.2 | XP_006254659.1 | 7 - 8 | 1104 | 76 | |
XM_008772043.1 | XP_008770265.1 | 8 - 9 | 1091 | 76 | |
GenBank mRNA | BC087108.1 | - | 7 - 8 | 929 | 76 |
FQ226177.1 | - | 3 - 4 | 280 | 76 |