The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012163.1 | NP_001012163.1 | 8 - 9 | 1290 | 64 |
XM_006254596.2 | XP_006254658.1 | 9 - 10 | 1196 | 64 | |
XM_006254597.2 | XP_006254659.1 | 9 - 10 | 1234 | 64 | |
XM_008772043.1 | XP_008770265.1 | 10 - 11 | 1221 | 64 | |
GenBank mRNA | BC087108.1 | - | 8 - 9 | 1290 | 64 |
FQ226177.1 | - | 5 - 6 | 410 | 64 |