Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sim2
Entrez Gene ID
304071
Gene Name
single-minded family bHLH transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.11: 34315611 - 34355181 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107108.3 NP_001100578.1 5 - 6 1101 69

View Full Product Details