The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013184.1 | NP_001013202.1 | 5 - 6 | 325 | 66 |
GenBank mRNA | BC086423.1 | - | 5 - 6 | 325 | 66 |
FQ220876.1 | - | 5 - 6 | 363 | 66 | |
FQ222121.1 | - | 5 - 6 | 364 | 66 | |
FQ230156.1 | - | 5 - 6 | 328 | 66 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751029.3 | XP_003751077.1 | 5 - 6 | 396 | 66 |