The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013429.1 | NP_001013447.1 | 1 - 2 | 401 | 60 |
GenBank mRNA | BC091387.1 | - | 1 - 2 | 401 | 60 |