Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt10
Entrez Gene ID
60567
Gene Name
synaptotagmin X
Gene Aliases
-
Chromosome Location
Chr.7: 130769039 - 130827030 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031666.1 NP_113854.1 3 - 4 1081 88
GenBank mRNA AF375463.1 - 3 - 4 1081 88
DQ437524.1 - 3 - 4 1081 88

View Full Product Details