The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024877.1 | NP_001020048.1 | 20 - 21 | 1817 | 63 |
XM_006242186.2 | XP_006242248.1 | 19 - 20 | 1782 | 63 | |
XM_006242187.2 | XP_006242249.1 | 19 - 20 | 1779 | 63 | |
XM_006242188.2 | XP_006242250.1 | 19 - 20 | 1743 | 63 | |
GenBank mRNA | BC097379.1 | - | 20 - 21 | 1817 | 63 |