The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085485.1 | NP_001078954.1 | 6 - 7 | 1612 | 58 |
GenBank mRNA | BC166588.1 | - | 6 - 7 | 1510 | 58 |
DV724467.1 | - | 2 - 3 | 418 | 58 | |
DV727162.1 | - | 3 - 4 | 515 | 58 |