Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prickle1
Entrez Gene ID
315259
Gene Name
prickle homolog 1 (Drosophila)
Gene Aliases
Rilp
Chromosome Location
Chr.7: 134702964 - 134799244 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199396.1 NP_955428.1 1 - 2 579 56

View Full Product Details