Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prickle1
Entrez Gene ID
315259
Gene Name
prickle homolog 1 (Drosophila)
Gene Aliases
Rilp
Chromosome Location
Chr.7: 134702964 - 134799244 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199396.1 NP_955428.1 6 - 7 1407 73
XM_006242213.2 XP_006242275.1 5 - 6 840 73
XM_006242214.2 XP_006242276.1 6 - 7 3304 73
XM_006242215.2 XP_006242277.1 6 - 7 1235 73
GenBank mRNA AF399843.1 - 3 - 4 596 73

View Full Product Details