Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gxylt1
Entrez Gene ID
300173
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
Glt8d3
Chromosome Location
Chr.7: 134447578 - 134495836 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100887.1 NP_001094357.1 1 - 2 254 122

View Full Product Details