The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031567.1 | NP_113755.1 | 7 - 8 | 1087 | 82 |
XM_006240850.2 | XP_006240912.1 | 7 - 8 | 1173 | 82 | |
GenBank mRNA | AB012233.2 | - | 7 - 8 | 1087 | 82 |